missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D6160 Aroclor 1260 35 μg/mL in Isooctane, Dr. Ehrenstorfer

Product Code. 30311352
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311352 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311352 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010471IO

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aroclor 1260
CAS 11096-82-5
Analyte or Component Names Aroclor 1260
Shipping Condition Room Temperature
Molecular Formula C12 H4 Cl6
Concentration 35 μg/mL
Certifications/Compliance ISO 17034
Synonym Aroclor 1260, PCB 1260
InChI Formula InChI=1S/C12H4Cl6/c13-7-3-1-5(9(15)11(7)17)6-2-4-8(14)12(18)10(6)16/h1-4H
SMILES ClC(C(Cl)=C(Cl)C=C1)=C1C2=C(Cl)C(Cl)=C(Cl)C=C2
IUPAC Name 1,2,3-trichloro-4-(2,3,4-trichlorophenyl)benzene
Solvent or Matrix Isooctane
Molecular Weight (g/mol) 360.87816
Quantity 1 mL
Formula Weight 357.8444 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.