missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM Method D5769 Quality Control Reference Material without IS in Isooctane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣2,2,4-Trimethylpentane∣Benzene∣Ethylbenzene∣m-Xylene (1,3-Dimethylbenzene)∣Naphthalene∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Octane∣o-Xylene (1,2-Dimethylbenzene)∣Toluene |
| CAS | 95-93-2, 95-63-6, 540-84-1, 71-43-2, 100-41-4, 108-38-3 |
| Analyte or Component Names | 1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣2,2,4-Trimethylpentane∣Benzene∣Ethylbenzene∣m-Xylene (1,3-Dimethylbenzene)∣Naphthalene∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Octane∣o-Xylene (1,2-Dimethylbenzene)∣Toluene |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Isooctane |
| Quantity | 10 x 2 mL |
| Concentration or Composition (by Analyte or Components) | m-Xylene (1,3-Dimethylbenzene) 3 wt%∣o-Xylene (1,2-Dimethylbenzene) 3 wt%∣n-Dodecane 5 wt%∣2,2,4-Trimethylpentane 12 wt%∣n-Decane 12 wt%∣n-Hexane 12 wt%∣n-Heptane 16.9 wt%∣n-Octane 16.9 wt%∣Benzene 10000 mg/kg∣Naphthalene 10000 mg/kg∣1,2,4,5-Tetramethylbenzene 20000 mg/kg∣1,2,4-Trimethylbenzene 30000 mg/kg∣Ethylbenzene 30000 mg/kg∣Toluene 90000 mg/kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Individuate un'opportunità di miglioramento?