missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D5769 Internal Standard Mixture, Dr. Ehrenstorfer

Product Code. 30309776
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Product Code. Quantity unitSize
30309776 10 mL 10mL
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30309776

Brand: Dr. Ehrenstorfer DREGA09000137

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzene-D6∣Ethylbenzene-d10∣Naphthalene D8∣Toluene D8
CAS 1076-43-3, 25837-05-2, 1146-65-2, 2037-26-5
Analyte or Component Names Benzene-D6∣Ethylbenzene-d10∣Naphthalene D8∣Toluene D8
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Quantity 10 mL
Concentration or Composition (by Analyte or Components) Benzene-D6 2 wt%∣Ethylbenzene-d10 2 wt%∣Naphthalene D8 9 wt%∣Toluene D8 58 wt%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.