missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D5769 Internal Standard Mixture, Dr. Ehrenstorfer

Product Code. 30321613
Change view
Click to view available options
Quantity:
5 x 10 mL
Unit Size:
50mL
Product Code. Quantity unitSize
30321613 5 x 10 mL 50mL
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30321613

Brand: Dr. Ehrenstorfer DREGS09000137

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzene-D6∣Ethylbenzene-d10∣Naphthalene D8∣Toluene D8
CAS 1076-43-3, 25837-05-2, 1146-65-2, 2037-26-5
Analyte or Component Names Benzene-D6∣Ethylbenzene-d10∣Naphthalene D8∣Toluene D8
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Quantity 5 x 10 mL
Concentration or Composition (by Analyte or Components) Benzene-D6 2 wt%∣Ethylbenzene-d10 2 wt%∣Naphthalene D8 8.8 wt%∣Toluene D8 57.9 wt%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.