missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D5191 Vapor Pressure - 68.3kPa (9.91 psi), Dr. Ehrenstorfer

Product Code. 30313618
Click to view available options
Quantity:
10 x 10 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 30313618

Brand: Dr. Ehrenstorfer DREGS09010494

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopentane
CAS 287-92-3
Analyte or Component Names Cyclopentane
Shipping Condition Room Temperature
Molecular Formula C5 H10
Concentration 100 %
Certifications/Compliance ISO 17034
Synonym Cyclopentane, Exxsol Cyclopentane S, Exxsol HP 95, H&N, IP 85, Marukazol FH, NSC 60213, Pentamethylene, Zeonsolv HP
InChI Formula InChI=1S/C5H10/c1-2-4-5-3-1/h1-5H2
SMILES C1CCCC1
IUPAC Name cyclopentane
Molecular Weight (g/mol) 70.13
Quantity 10 x 10 mL
Formula Weight 70.0783 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.