missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D5191 Vapor Pressure - 51.1kPa (7.41 psi), Dr. Ehrenstorfer

Product Code. 30319101
Change view
Click to view available options
Quantity:
10 x 10 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319101 10 x 10 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319101 Supplier Dr. Ehrenstorfer Supplier No. DREGS09010493
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 2,3-Dimethylbutane
CAS 79-29-8
Analyte or Component Names 2,3-Dimethylbutane
Shipping Condition Room Temperature
Molecular Formula C6 H14
Concentration 100 %
Certifications/Compliance ISO 17034
Synonym Butane, 2,3-dimethyl-, 2,3-Dimethylbutane, 1,1,2,2-Tetramethylethane, Biisopropyl, Diisopropyl, NSC 24837
InChI Formula InChI=1S/C6H14/c1-5(2)6(3)4/h5-6H,1-4H3
SMILES CC(C)C(C)C
IUPAC Name 2,3-dimethylbutane
Molecular Weight (g/mol) 86.18
Quantity 10 x 10 mL
Formula Weight 86.1096 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.