missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D5191 Vapor Pressure - 46.7 kPa (6.77 psi), Dr. Ehrenstorfer

Product Code. 30316532
Change view
Click to view available options
Quantity:
10 x 10 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316532 10 x 10 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316532 Supplier Dr. Ehrenstorfer Supplier No. DREGS09010492
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methylpentane
CAS 107-83-5
Analyte or Component Names 2-Methylpentane
Shipping Condition Room Temperature
Molecular Formula C6 H14
Concentration 100 %
Certifications/Compliance ISO 17034
Synonym Pentane, 2-methyl-, 2-Methylpentane, 1,1-Dimethylbutane, Isohexane, Kyowasol C 600M, Kyowazol C 600, NSC 66496
InChI Formula InChI=1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3
SMILES CCCC(C)C
IUPAC Name 2-methylpentane
Molecular Weight (g/mol) 86.18
Quantity 10 x 10 mL
Formula Weight 86.1096 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.