missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D5134 Linearity Check Mixture, Dr. Ehrenstorfer

Product Code. 30311257
Click to view available options
Quantity:
10 x 25 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30311257

Brand: Dr. Ehrenstorfer DREGS0900107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-dimethylheptane∣2,4-Dimethylhexane∣2-Methylheptane∣2-Methylhexane∣Benzene∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣Toluene
CAS 2213-23-2, 589-43-5, 592-27-8, 591-76-4, 71-43-2, 142-82-5
Analyte or Component Names 2,4-dimethylheptane∣2,4-Dimethylhexane∣2-Methylheptane∣2-Methylhexane∣Benzene∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣Toluene
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Quantity 10 x 25 mg
Concentration or Composition (by Analyte or Components) 2,4-dimethylheptane 10 %∣2,4-Dimethylhexane 10 %∣2-Methylheptane 10 %∣2-Methylhexane 10 %∣Benzene 10 %∣n-Heptane 10 %∣n-Hexane 10 %∣n-Nonane 10 %∣n-Octane 10 %∣Toluene 10 %
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.