missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM Method D5134 Column Evaluation Mixture, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 2,3,3-trimethylpentane∣2-Methylheptane∣2-Methylpentane∣4-Methylheptane∣n-Heptane∣n-Octane∣Toluene |
| CAS | 560-21-4, 592-27-8, 107-83-5, 589-53-7, 142-82-5, 111-65-9 |
| Analyte or Component Names | 2,3,3-trimethylpentane∣2-Methylheptane∣2-Methylpentane∣4-Methylheptane∣n-Heptane∣n-Octane∣Toluene |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Toluene 0.5 %∣2,3,3-trimethylpentane 1 %∣2-Methylheptane 1 %∣4-Methylheptane 1 %∣n-Heptane 1 %∣n-Octane 1 %∣2-Methylpentane 94.5 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction