missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM Method D4059 Aroclor 1260 500 μg/g in Transformer Oil, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Aroclor 1260 |
| CAS | 11096-82-5 |
| Analyte or Component Names | Aroclor 1260 |
| Shipping Condition | +20°C |
| Molecular Formula | C12 H4 Cl6 |
| Concentration | 500 μg/g |
| Certifications/Compliance | ISO 17034 |
| Synonym | Aroclor 1260, PCB 1260 |
| InChI Formula | InChI=1S/C12H4Cl6/c13-7-3-1-5(9(15)11(7)17)6-2-4-8(14)12(18)10(6)16/h1-4H |
| SMILES | ClC(C(Cl)=C(Cl)C=C1)=C1C2=C(Cl)C(Cl)=C(Cl)C=C2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?