missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D4059 Aroclor 1254 500 μg/g in Transformer Oil, Dr. Ehrenstorfer

Product Code. 30323766
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323766 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323766 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010447TR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aroclor 1254
CAS 11097-69-1
Analyte or Component Names Aroclor 1254
Shipping Condition +20°C
Concentration 50 μg/g
Certifications/Compliance ISO 17034
Synonym Aroclor 1254, PCB 1254
Solvent or Matrix Transformer Oil
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.