missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM Method D4059 Aroclor 1254 50 μg/g in Transformer Oil, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Aroclor 1254 |
| CAS | 11097-69-1 |
| Analyte or Component Names | Aroclor 1254 |
| Shipping Condition | +20°C |
| Concentration | 500 μg/g |
| Certifications/Compliance | ISO 17034 |
| Synonym | Aroclor 1254, PCB 1254 |
| Solvent or Matrix | Transformer Oil |
| Quantity | 5 x 1 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?