missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D3710 Quantitative Calibration Standard, Dr. Ehrenstorfer

Product Code. 30320481
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320481 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320481 Supplier Dr. Ehrenstorfer Supplier No. DREGA0900105
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dimethylpentane∣2-Methylbutane∣2-Methylpentane∣n-Butylbenzene∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Octane∣n-Pentadecane∣n-Pentane∣n-Tetradecane∣n-Tridecane∣Propylbenzene∣p-Xylene (1,4-Dimethylbenzene)∣Toluene
CAS 108-08-7, 78-78-4, 107-83-5, 104-51-8, 124-18-5, 112-40-3
Analyte or Component Names 2,4-Dimethylpentane∣2-Methylbutane∣2-Methylpentane∣n-Butylbenzene∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Octane∣n-Pentadecane∣n-Pentane∣n-Tetradecane∣n-Tridecane∣Propylbenzene∣p-Xylene (1,4-Dimethylbenzene)∣Toluene
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Quantity 1 mL
Concentration or Composition (by Analyte or Components) n-Pentadecane 2.3 %∣n-Tetradecane 2.3 %∣n-Tridecane 2.3 %∣n-Butylbenzene 3.5 %∣n-Decane 3.5 %∣n-Dodecane 3.5 %∣Propylbenzene 4.7 %∣2,4-Dimethylpentane 5.8 %∣2-Methylpentane 5.8 %∣n-Hexane 5.8 %∣n-Octane 5.8 %∣n-Pentane 8.1 %∣2-Methylbutane 10.5 %∣n-Heptane 10.5 %∣Toluene 11.6 %∣p-Xylene (1,4-Dimethylbenzene) 14 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.