missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D3606 Benzene in Gasoline Kit with IS in Isooctane, Dr. Ehrenstorfer

Product Code. 30320621
Change view
Click to view available options
Quantity:
7 x 2 mL
Unit Size:
14mL
Product Code. Quantity unitSize
30320621 7 x 2 mL 14mL
1 options
This item is not returnable. View return policy

Product Code. 30320621

Brand: Dr. Ehrenstorfer DREGK09000100IO

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Butanol∣Benzene∣Toluene
CAS 78-92-2, 71-43-2, 108-88-3
Analyte or Component Names 2-Butanol∣Benzene∣Toluene
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Isooctane
Quantity 7 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.