missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM Method D2887 Hydrocarbon Window Defining Solution, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Heneicosane∣n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hentriacontane∣n-Heptacosane∣n-Heptadecane∣n-Heptatriacontane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Nonacosane∣n-Nonadecane∣n-Nonane∣n-Nonatriacontane∣n-Octadecane∣n-Octane∣n-Octatriacontane∣n-Pentacosane∣n-Pentadecane∣n-Pentatriacontane∣n-Tetradecane∣n-Tetratriacontane∣n-Tricosane∣n-Tridecane∣n-Tritriacontane∣n-Undecane∣Octacosane∣Phytane∣Pristane∣Tetracontane∣Tetracosane∣Triacontane |
| CAS | 629-94-7, 124-18-5, 629-97-0, 112-40-3, 544-85-4, 112-95-8 |
| Analyte or Component Names | Heneicosane∣n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hentriacontane∣n-Heptacosane∣n-Heptadecane∣n-Heptatriacontane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Nonacosane∣n-Nonadecane∣n-Nonane∣n-Nonatriacontane∣n-Octadecane∣n-Octane∣n-Octatriacontane∣n-Pentacosane∣n-Pentadecane∣n-Pentatriacontane∣n-Tetradecane∣n-Tetratriacontane∣n-Tricosane∣n-Tridecane∣n-Tritriacontane∣n-Undecane∣Octacosane∣Phytane∣Pristane∣Tetracontane∣Tetracosane∣Triacontane |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 82.5% Chloroform/12.5% Carbon Disulfide/5% Hexane |
| Quantity | 5 x 1 mL |
| Concentration or Composition (by Analyte or Components) | Heneicosane 500 μg/mL∣n-Decane 500 μg/mL∣n-Docosane 500 μg/mL∣n-Dodecane 500 μg/mL∣n-Dotriacontane 500 μg/mL∣n-Eicosane 500 μg/mL∣n-Hentriacontane 500 μg/mL∣n-Heptacosane 500 μg/mL∣n-Heptadecane 500 μg/mL∣n-Heptatriacontane 500 μg/mL∣n-Hexacosane 500 μg/mL∣n-Hexadecane 500 μg/mL∣n-Hexatriacontane 500 μg/mL∣n-Nonacosane 500 μg/mL∣n-Nonadecane 500 μg/mL∣n-Nonane 500 μg/mL∣n-Nonatriacontane 500 μg/mL∣n-Octadecane 500 μg/mL∣n-Octane 500 μg/mL∣n-Octatriacontane 500 μg/mL∣n-Pentacosane 500 μg/mL∣n-Pentadecane 500 μg/mL∣n-Pentatriacontane 500 μg/mL∣n-Tetradecane 500 μg/mL∣n-Tetratriacontane 500 μg/mL∣n-Tricosane 500 μg/mL∣n-Tridecane 500 μg/mL∣n-Tritriacontane 500 μg/mL∣n-Undecane 500 μg/mL∣Octacosane 500 μg/mL∣Phytane 500 μg/mL∣Pristane 500 μg/mL∣Tetracontane 500 μg/mL∣Tetracosane 500 μg/mL∣Triacontane 500 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?