missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D2887 Calibration Solution, Dr. Ehrenstorfer

Product Code. 30312244
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312244 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312244 Supplier Dr. Ehrenstorfer Supplier No. DREGA0900101CH
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Decane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Heptane∣n-Hexadecane∣n-Hexane∣n-Hexatriacontane∣n-Nonane∣n-Octadecane∣n-Octane∣n-Tetradecane∣n-Tetratetracontane∣n-Undecane∣Octacosane∣Tetracontane∣Tetracosane
CAS 124-18-5, 112-40-3, 544-85-4, 112-95-8, 142-82-5, 544-76-3
Analyte or Component Names n-Decane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Heptane∣n-Hexadecane∣n-Hexane∣n-Hexatriacontane∣n-Nonane∣n-Octadecane∣n-Octane∣n-Tetradecane∣n-Tetratetracontane∣n-Undecane∣Octacosane∣Tetracontane∣Tetracosane
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Chloroform
Quantity 1 mL
Concentration or Composition (by Analyte or Components) n-Dotriacontane 100 μg/mL∣n-Hexatriacontane 100 μg/mL∣n-Tetratetracontane 100 μg/mL∣Octacosane 100 μg/mL∣Tetracontane 100 μg/mL∣n-Eicosane 200 μg/mL∣Tetracosane 200 μg/mL∣n-Octadecane 500 μg/mL∣n-Heptane 600 μg/mL∣n-Hexane 600 μg/mL∣n-Nonane 800 μg/mL∣n-Octane 800 μg/mL∣n-Hexadecane 1000 μg/mL∣n-Decane 1200 μg/mL∣n-Dodecane 1200 μg/mL∣n-Tetradecane 1200 μg/mL∣n-Undecane 1200 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.