missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D1319 Aromatics and Olefins by FIA Mixture 20% LV Toluene and 5% LV 1-Hexene in Isooctane, Dr. Ehrenstorfer

Product Code. 30320558
Change view
Click to view available options
Quantity:
16oz
Unit Size:
Pack of 16
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320558 16oz Pack of 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320558 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000379IO

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Hexene∣2,2,4-Trimethylpentane∣Toluene
CAS 592-41-6, 540-84-1, 108-88-3
Analyte or Component Names 1-Hexene∣2,2,4-Trimethylpentane∣Toluene
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Isooctane
Quantity 16oz
Concentration or Composition (by Analyte or Components) 1-Hexene 5 wt%∣Toluene 20 wt%∣2,2,4-Trimethylpentane 75 wt%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.