missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM D8399 Pesticide Mixture 4 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Acequinocyl∣Aldicarb∣Bifenazate∣Clofentezine∣Daminozide∣Dinotefuran∣Dodemorph∣Methoprene∣Pyrethrins∣Spinosad∣Thiophanate-methyl |
| CAS | 57960-19-7, 116-06-3, 149877-41-8, 74115-24-5, 1596-84-5, 165252-70-0 |
| Analyte or Component Names | Acequinocyl∣Aldicarb∣Bifenazate∣Clofentezine∣Daminozide∣Dinotefuran∣Dodemorph∣Methoprene∣Pyrethrins∣Spinosad∣Thiophanate-methyl |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Acequinocyl 100 μg/mL∣Aldicarb 100 μg/mL∣Bifenazate 100 μg/mL∣Clofentezine 100 μg/mL∣Daminozide 100 μg/mL∣Dinotefuran 100 μg/mL∣Dodemorph 100 μg/mL∣Methoprene 100 μg/mL∣Pyrethrins 100 μg/mL∣Spinosad 100 μg/mL∣Thiophanate-methyl 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction