missing translation for 'onlineSavingsMsg'
Learn More

ASTM D8399 Pesticide Mixture 3 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30318497
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30318497

Brand: Dr. Ehrenstorfer DREA50000734AL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azadirachtin A∣Benzovindiflupyr∣Clothianidin∣Cyantraniliprole∣Deltamethrin∣Fenoxycarb∣Fenpyroximate (E/Z Mix)∣Fenvalerate∣Fludioxonil∣Fluopyram∣Mevinphos∣Permethrin
CAS 11141-17-6, 1072957-71-1, 210880-92-5, 736994-63-1, 52918-63-5, 72490-01-8
Analyte or Component Names Azadirachtin A∣Benzovindiflupyr∣Clothianidin∣Cyantraniliprole∣Deltamethrin∣Fenoxycarb∣Fenpyroximate (E/Z Mix)∣Fenvalerate∣Fludioxonil∣Fluopyram∣Mevinphos∣Permethrin
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Azadirachtin A 100 μg/mL∣Benzovindiflupyr 100 μg/mL∣Clothianidin 100 μg/mL∣Cyantraniliprole 100 μg/mL∣Deltamethrin 100 μg/mL∣Fenoxycarb 100 μg/mL∣Fenpyroximate (E/Z Mix) 100 μg/mL∣Fenvalerate 100 μg/mL∣Fludioxonil 100 μg/mL∣Fluopyram 100 μg/mL∣Mevinphos 100 μg/mL∣Permethrin 100 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.