missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM D8399 Pesticide Mixture 2 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Abamectin∣Acephate∣Allethrin∣Bifenthrin∣Chlorpyrifos∣Coumaphos∣Diazinon∣Dichlorvos∣Dimethoate∣Ethoprophos∣Fensulfothion∣Fenthion∣Fipronil∣Kresoxim-methyl∣Metalaxyl∣Methomyl∣Oxamyl∣Paclobutrazol∣Phenothrin∣Prallethrin∣Propiconazole∣Propoxur∣Pyraclostrobin∣Pyridaben∣Resmethrin∣Spirotetramat∣Teflubenzuron∣Tetramethrin |
| CAS | 71751-41-2, 30560-19-1, 584-79-2, 82657-04-3, 2921-88-2, 56-72-4 |
| Analyte or Component Names | Abamectin∣Acephate∣Allethrin∣Bifenthrin∣Chlorpyrifos∣Coumaphos∣Diazinon∣Dichlorvos∣Dimethoate∣Ethoprophos∣Fensulfothion∣Fenthion∣Fipronil∣Kresoxim-methyl∣Metalaxyl∣Methomyl∣Oxamyl∣Paclobutrazol∣Phenothrin∣Prallethrin∣Propiconazole∣Propoxur∣Pyraclostrobin∣Pyridaben∣Resmethrin∣Spirotetramat∣Teflubenzuron∣Tetramethrin |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Abamectin 100 μg/mL∣Acephate 100 μg/mL∣Allethrin 100 μg/mL∣Bifenthrin 100 μg/mL∣Chlorpyrifos 100 μg/mL∣Coumaphos 100 μg/mL∣Diazinon 100 μg/mL∣Dichlorvos 100 μg/mL∣Dimethoate 100 μg/mL∣Ethoprophos 100 μg/mL∣Fensulfothion 100 μg/mL∣Fenthion 100 μg/mL∣Fipronil 100 μg/mL∣Kresoxim-methyl 100 μg/mL∣Metalaxyl 100 μg/mL∣Methomyl 100 μg/mL∣Oxamyl 100 μg/mL∣Paclobutrazol 100 μg/mL∣Phenothrin 100 μg/mL∣Prallethrin 100 μg/mL∣Propiconazole 100 μg/mL∣Propoxur 100 μg/mL∣Pyraclostrobin 100 μg/mL∣Pyridaben 100 μg/mL∣Resmethrin 100 μg/mL∣Spirotetramat 100 μg/mL∣Teflubenzuron 100 μg/mL∣Tetramethrin 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction