missing translation for 'onlineSavingsMsg'
Learn More

ASTM D8399 Pesticide Mixture 1 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30309074
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30309074

Brand: Dr. Ehrenstorfer DREA50000732AL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetamiprid∣Azoxystrobin∣Boscalid∣Buprofezin∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Cyprodinil∣Dimethomorph∣Etofenprox∣Etoxazole∣Flonicamid∣Hexythiazox∣Imazalil∣Imidacloprid∣Iprodione∣Malathion∣Methiocarb∣Myclobutanil∣Novaluron∣Phosmet∣Piperonyl butoxide∣Pirimicarb∣Spinetoram∣Spiroxamine∣Tebuconazole∣Tebufenozide∣Tetrachlorvinphos∣Thiacloprid∣Thiamethoxam∣Trifloxystrobin
CAS 135410-20-7, 131860-33-8, 188425-85-6, 69327-76-0, 63-25-2, 1563-66-2
Analyte or Component Names Acetamiprid∣Azoxystrobin∣Boscalid∣Buprofezin∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Cyprodinil∣Dimethomorph∣Etofenprox∣Etoxazole∣Flonicamid∣Hexythiazox∣Imazalil∣Imidacloprid∣Iprodione∣Malathion∣Methiocarb∣Myclobutanil∣Novaluron∣Phosmet∣Piperonyl butoxide∣Pirimicarb∣Spinetoram∣Spiroxamine∣Tebuconazole∣Tebufenozide∣Tetrachlorvinphos∣Thiacloprid∣Thiamethoxam∣Trifloxystrobin
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Acetamiprid 100 μg/mL∣Azoxystrobin 100 μg/mL∣Boscalid 100 μg/mL∣Buprofezin 100 μg/mL∣Carbaryl 100 μg/mL∣Carbofuran 100 μg/mL∣Chlorantraniliprole 100 μg/mL∣Cyprodinil 100 μg/mL∣Dimethomorph 100 μg/mL∣Etofenprox 100 μg/mL∣Etoxazole 100 μg/mL∣Flonicamid 100 μg/mL∣Hexythiazox 100 μg/mL∣Imazalil 100 μg/mL∣Imidacloprid 100 μg/mL∣Iprodione 100 μg/mL∣Malathion 100 μg/mL∣Methiocarb 100 μg/mL∣Myclobutanil 100 μg/mL∣Novaluron 100 μg/mL∣Phosmet 100 μg/mL∣Piperonyl butoxide 100 μg/mL∣Pirimicarb 100 μg/mL∣Spinetoram 100 μg/mL∣Spiroxamine 100 μg/mL∣Tebuconazole 100 μg/mL∣Tebufenozide 100 μg/mL∣Tetrachlorvinphos 100 μg/mL∣Thiacloprid 100 μg/mL∣Thiamethoxam 100 μg/mL∣Trifloxystrobin 100 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.