missing translation for 'onlineSavingsMsg'
Learn More

ASTM D4294, Sulfur Set B in Light Oil (20 cSt): S @ 1000, 2500, 5000, 10000 μg/g, VHG™

Product Code. 17962976
Change view
Click to view available options
Quantity:
4 x 100 mL
Unit Size:
400mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17962976 4 x 100 mL 400mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17962976 Supplier VHG Labs Supplier No. VHGKTS20MIN4294B4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ASTM D4294, Sulfur Set B in Light Oil (20 cSt): S @ 1000, 2500, 5000, 10000 μg/g
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfur
CAS 7704-34-9
Methods XRF/UVF
Analyte or Component Names Sulfur
Shipping Condition Room Temperature
Molecular Formula S
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/S
SMILES [S]
IUPAC Name sulfur
Solvent or Matrix 20 cSt Oil
Molecular Weight (g/mol) 32.065
Quantity 4 x 100 mL
Formula Weight 31.972 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.