missing translation for 'onlineSavingsMsg'
Learn More

ASTM D4294, Curve #2, VHG™

Product Code. 17981886
Change view
Click to view available options
Quantity:
5 x 100 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981886 5 x 100 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17981886 Supplier VHG Labs Supplier No. ALKKTSMINW6CRV2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Se returpolicy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ASTM D4294, Curve #2
TRUSTED_SUSTAINABILITY

Specifikationer

Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 75 cSt Mineral Oil
Quantity 5 x 100 mL
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.