missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASTL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5149481
This item is not returnable.
View return policy
Description
ASTL Polyclonal Antibody for Western Blot
Specifications
| ASTL | |
| Polyclonal | |
| Unconjugated | |
| ASTL | |
| astacin-like metalloendopeptidase; astacin-like metalloendopeptidase (M12 family); astacin-like metallo-endopeptidase (M12 family); ASTL; Oocyte astacin; ovastacin; SAS1B; sperm acrosomal SLLP1 binding | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 431705 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| Conc. Not Determined | |
| PBS with 0.1% BSA, 50% glycerol and 0.09% sodium azide | |
| Q6HA08 | |
| ASTL | |
| Mouse myeloma cell line NS0-derived recombinant human Ovastacin, Ala24-Asp431. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction