Learn More
Invitrogen™ Astegolimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559096
 View more versions of this product
Product Code. 30282556
  • £1002.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Astegolimab is a biosimilar that targets IL1RL1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Astegolimab
Recombinant Monoclonal
Unconjugated
AMG-282; MSTT-1041A; RG-6149; RO-7187807
Protein A
RUO
Human
Antibody
IgG2SA
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized