missing translation for 'onlineSavingsMsg'
Learn More

Astaxanthin, TRC

Product Code. 17979214
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17979214 50 mg 50mg
17959784 10 mg 10mg
17938154 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17979214 Supplier Toronto Research Chemicals Supplier No. TRCA79002550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 472-61-7
Molecular Formula C40 H52 O4
Molecular Weight (g/mol) 596.84
Synonym Astaxanthin (6CI), β-Carotene-4,4'-dione, 3,3'-dihydroxy-, all-trans- (8CI), (3S,3'S)-3,3'-Dihydroxy-β,β-carotene-4,4'-dione, (3S,3'S)-Astaxanthin, (3S,3'S)-Astaxanthin, (3S,3'S)-all-trans-Astaxanthin, (S,S)-Astaxanthin, Aquasta, AstaREAL, AstaREAL A1010, AstaXin, Astabio AR 1, Astabio AR 5, Astacarox, Astareal ACT, Astared, Astax S, Astaxanthin, all-trans-, Astots 10O, Astots 5O, Astots ST, BioAstin, BioAstin oleoresin, Carophyll Pink, Lucantin Pink, NatuRose, Natupink, Ovoester, Puresta W 05, all-trans-(3S,3'S)-Astaxanthin, all-trans-Astaxanthin, trans-Astaxanthin
IUPAC Name (6S)-6-hydroxy-3-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-18-[(4S)-4-hydroxy-2,6,6-trimethyl-3-oxocyclohexen-1-yl]-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaenyl]-2,4,4-trimethylcyclohex-2-en-1-one
SMILES C\C(=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C(=O)[C@@H](O)CC1(C)C)\C=C\C=C(/C)\C=C\C2=C(C)C(=O)[C@@H](O)CC2(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C40H52O4/c1-27(17-13-19-29(3)21-23-33-31(5)37(43)35(41)25-39(33,7)8)15-11-12-16-28(2)18-14-20-30(4)22-24-34-32(6)38(44)36(42)26-40(34,9)10/h11-24,35-36,41-42H,25-26H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,27-15+,28-16+,29-19+,30-20+/t35-,36-/m0/s1
Formula Weight 596.3866
Chemical Name or Material Astaxanthin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.