Learn More
Karl Hecht™ Assisent™ Polyethylene Pasteur Pipets
Description
- Manufactured from high quality, chemically inert, non-toxic, low-density polyethylene
- Excellent for transferring samples to sample cups and for drop counting
- Disposable
Specifications
Specifications
| Material | Polyethylene |
| Volume (Metric) | 3 mL |
| Type | Pasteur Pipet |
| Disposable | Single-use |