missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX AssayComplete™ Protein Dilution Buffer
Click to view available options
Quantity:
2 x 50 mL
Unit Size:
100mL
Description
AssayComplete Protein Dilution Buffer is a specially formulated dilution buffer designed for use in reconstituting protein agonists and diluting other ligands (antibodies, small molecules, etc) to the proper stock concentration for use in DiscoverX cell-based assays. This product is provided as 2 bottles of 250mL buffer each.
Specifications
Specifications
| Chemical Name or Material | Buffer |
| For Use With (Application) | Dilution Buffer |
| Quantity | 2 x 50 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction