missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nabertherm Assay Furnace, 1300°C
Shop All Nabertherm Products
Click to view available options
Capacity (Metric):
8 L
25 L
Unit Size:
Each
Description
- Muffle heated from four sides (three sides for assay furnace N 25/13 CUP)
- Heating elements and insulation protected by muffle
- Simple replacement of muffle
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Warm furnace can be opened
- Tool holder on furnace
- Stainless steel exhaust chimney above the door opening for connection of an exhaust system
- Dual shell housing with fan cooling to reduce exterior temperatures
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications
Specifications
| Capacity (Metric) | 8 L |
| Temperature (Metric) | 1300°C |
| Dimensions (D x W x H) Interior | 340 x 260 x 95 mm |
| Dimensions (L x W x H) Exterior | 1335 x 900 x 2100 mm |
| Weight (Metric) | 510 kg |
| Electrical Requirements | 3-phase |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction