missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm Assay Furnace, 1300°C

Product Code. 15928270 Shop All Nabertherm Products
Change view
Click to view available options
Capacity (Metric):
8 L
25 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15928270 8 L Each
15938270 25 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15928270 Supplier Nabertherm Supplier No. 001490400

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used primarily for the assay of precious metals and ashing processes where the insulation and heating must be protected from emerging gasses and vapours. The assay furnace is equipped with an integrated exhaust hood above the furnace door for connection to and exhaust system.

  • Muffle heated from four sides (three sides for assay furnace N 25/13 CUP)
  • Heating elements and insulation protected by muffle
  • Simple replacement of muffle
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Warm furnace can be opened
  • Tool holder on furnace
  • Stainless steel exhaust chimney above the door opening for connection of an exhaust system
  • Dual shell housing with fan cooling to reduce exterior temperatures
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive

Specifications

Capacity (Metric) 8 L
Temperature (Metric) 1300°C
Dimensions (D x W x H) Interior 340 x 260 x 95 mm
Dimensions (L x W x H) Exterior 1335 x 900 x 2100 mm
Weight (Metric) 510 kg
Electrical Requirements 3-phase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.