missing translation for 'onlineSavingsMsg'
Learn More

ASRGL1 Mouse, Clone: CRASH/1289, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 80150-MSM1-P1

 View more versions of this product

Product Code. 15991118

  • £423.29 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

ASRGL1
Cytoplasmic and Nuclear
CRASH/1289
Unconjugated
XP_005274363
Mouse
17-25kDa
Protein A/G Purified
RUO
80150
IgG1
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
ASRGL1
Recombinant human full-length ASRGL1
HeLa cells. Fallopian Tube, Endometrium, Uterus or Cervix.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.