missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASRGL1 Mouse, Clone: CRASH/1289, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 80150-MSM1-P0
This item is not returnable.
View return policy
Specifications
| ASRGL1 | |
| Cytoplasmic and Nuclear | |
| CRASH/1289 | |
| Unconjugated | |
| XP_005274363 | |
| Mouse | |
| 17-25kDa | |
| Protein A/G Purified | |
| RUO | |
| 80150 | |
| IgG1 |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| ASRGL1 | |
| Recombinant human full-length ASRGL1 | |
| HeLa cells. Fallopian Tube, Endometrium, Uterus or Cervix. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction