missing translation for 'onlineSavingsMsg'
Learn More

ASRGL1 Mouse anti-Human, Clone: CRASH/1290, Novus Biologicals™

Product Code. 18396104 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396104 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396104 Supplier Novus Biologicals Supplier No. NBP30736820UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ASRGL1 Monoclonal specifically detects ASRGL1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASRGL1
Applications ELISA
Classification Monoclonal
Clone CRASH/1290
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA
Formulation 10 mM PBS with 0.05% BSA
Gene Alias ALP1, ALPL-asparagine amidohydrolase, asparaginase like 1, asparaginase-like 1 protein, Asparaginase-like protein 1, CRASH, EC 3.5.1.1, FLJ22316, L-asparaginase
Host Species Mouse
Immunogen Recombinant human ASRGL1 fragment
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80150
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.