missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASRGL1 Mouse anti-Human, Clone-CRASH/1289, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54450-100UG
This item is not returnable.
View return policy
Description
ASRGL1 Monoclonal specifically detects ASRGL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ASRGL1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| ALP1, ALPL-asparagine amidohydrolase, asparaginase like 1, asparaginase-like 1 protein, Asparaginase-like protein 1, CRASH, EC 3.5.1.1, FLJ22316, L-asparaginase | |
| Mouse | |
| 21 kDa | |
| 100 μg | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| CRASH/1289 | |
| Unconjugated | |
| PBS with No Preservative | |
| ASRGL1 | |
| Recombinant human full-length ASRGL1 | |
| Protein A or G purified | |
| RUO | |
| 80150 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction