missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASPRV1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557071
This item is not returnable.
View return policy
Description
Immunogen sequence: SHLPKEIVFA NSMGKGYYLK GKIGKVPVRF LVDSGAQVSV VHPNLWEEVT DGDLDTLQPF ENVVKVANGA EMKILGVWDT AVSL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 25%.
Filaggrin is a structural protein that is crucial for in the development and maintenance of the skin barrier. This gene encodes a retroviral-like protease involved in profilaggrin-to-filaggrin processing. Expression is found primarily in the epidermis and inner root sheath of hair follicles.
Specifications
| ASPRV1 | |
| Polyclonal | |
| Unconjugated | |
| ASPRV1 | |
| 2300003P22Rik; AA986851; aspartic peptidase retroviral like 1; aspartic peptidase, retroviral-like 1; asprv1; LOW QUALITY PROTEIN: retroviral-like aspartic protease 1; MUNO; Retroviral-like aspartic protease 1; RGD1560859; SASP; SASPase; skin aspartic protease; skin-specific retroviral-like aspartic protease; TAPS; TPA-inducible aspartic proteinase-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 151516 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53RT3 | |
| ASPRV1 | |
| Recombinant protein corresponding to Human ASPRV1. Recombinant protein control fragment (Product #RP-96210). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction