missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Asprosin Monoclonal specifically detects Asprosin in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | Asprosin |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | Birdy-1 |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Gene Alias | FBN1, fibrillin 1 |
| Host Species | Mouse |
| Immunogen | Recombinant human asprosin. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?