missing translation for 'onlineSavingsMsg'
Learn More

Asprosin Antibody (Birdy-1), DyLight 350, Novus Biologicals™

Product Code. 18348152 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348152 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348152 Supplier Novus Biologicals Supplier No. NBP311677UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Asprosin Monoclonal specifically detects Asprosin in Human, Mouse samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Asprosin
Applications Western Blot, ELISA
Classification Monoclonal
Clone Birdy-1
Conjugate DyLight 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias FBN1, fibrillin 1
Host Species Mouse
Immunogen Recombinant human asprosin.
Purification Method >95%, by SDS-PAGE
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2200
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.