missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Asporin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112364
This item is not returnable.
View return policy
Description
Asporin Polyclonal Antibody for Western Blot, IHC (P), ELISA
ASPN belongs to a family of leucine-rich repeat (LRR) proteins associated with the cartilage matrix. The name asporin reflects the unique aspartate-rich N terminus and the overall similarity to decorin.
Specifications
| Asporin | |
| Polyclonal | |
| Unconjugated | |
| Aspn | |
| 4631401G09Rik; AA986886; ASPN; Asporin; asporin (LRR class 1); asporin proteoglycan; FLJ20129; OS3; periodontal ligament associated protein 1; periodontal ligament-associated protein 1; PLAP1; PLAP-1; SLRR1C; small leucine-rich protein 1C; UNQ215/PRO241 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 54829, 66695 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q99MQ4, Q9BXN1 | |
| Aspn | |
| Recombinant Human Asporin protein (33-380AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction