missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Asporin Polyclonal Antibody

Description
This antibody is predicted to react with rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
ASPN belongs to a family of leucine-rich repeat (LRR) proteins associated with the cartilage matrix. The name asporin reflects the unique aspartate-rich N terminus and the overall similarity to decorin.
Specifications
Specifications
| Antigen | Asporin |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Aspn |
| Gene Accession No. | Q99MQ4, Q9BXN1 |
| Gene Alias | 4631401G09Rik; AA986886; ASPN; Asporin; asporin (LRR class 1); asporin proteoglycan; FLJ20129; OS3; periodontal ligament associated protein 1; periodontal ligament-associated protein 1; PLAP1; PLAP-1; SLRR1C; small leucine-rich protein 1C; UNQ215/PRO241 |
| Gene Symbols | Aspn |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?