missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPM Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92395-0.02ml
This item is not returnable.
View return policy
Description
ASPM Polyclonal antibody specifically detects ASPM in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ASPM | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| ASP, asp (abnormal spindle) homolog, microcephaly associated (Drosophila), Asp homolog, Calmbp1, FLJ43117, MCPH5DKFZp686N06184, microcephaly associated (Drosophila), microcephaly, primary autosomal recessive 5 | |
| A synthetic peptide corresponding to a sequence within amino acids 450-550 of human ASPM (NP_060606.3). IFEELVEMKSNYYSFIKQNNPKFSAVQDISSHSHNKQPKRRPILSATVTKRKATCTRENQTEINKPKAKRCLNSAVGEHEKVINNQKEKEDFHSYLPIIDP | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 259266 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction