missing translation for 'onlineSavingsMsg'
Learn More

Aspirin, MedChemExpress

Product Code. 16481429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481429 1 g 1g
16438727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481429 Supplier Medchem Express Supplier No. HY14654
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aspirin (Acetylsalicylic Acid) is an orally active, potent and irreversible inhibitor of cyclooxygenase COX-1 and COX-2, with IC50 values of 5 and 210 μg/mL, respectively. Aspirin induces apoptosis. Aspirin inhibits the activation of NF-κB. Aspirin also inhibits platelet prostaglandin synthetase, and can prevent coronary artery and cerebrovascular thrombosis.

Aspirin (Acetylsalicylic Acid) is an orally active, potent and irreversible inhibitor of cyclooxygenase COX-1 and COX-2, with IC50 values of 5 and 210 μg/mL, respectively. Aspirin induces apoptosis. Aspirin inhibits the activation of NF-κB. Aspirin also inhibits platelet prostaglandin synthetase, and can prevent coronary artery and cerebrovascular thrombosis.

Specifications

Chemical Name or Material Aspirin
CAS 50-78-2
Color White
Purity Grade Notes Research
Molecular Formula C9H8O4
Quantity 1 g
Synonym Acetylsalicylic Acid ASA
Solubility Information DMSO : 400 mg/mL (2220.25 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.56 mM; Need ultrasonic)
SMILES OC(C1=C(OC(C)=O)C=CC=C1)=O
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.