missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ HDPE, Aspirator Bottle
Closure Material: Polypropylene; Color: White
Brand: BRAND™ 131174
This item is not returnable.
View return policy
Description
- Hight Density Polyethylene (HDPE)
- Narrow neck
- Includes polyprpylene stopcock, screw cap and carrying handle
Specifications
2.6 gal. | |
Polypropylene | |
45 mm | |
HDPE | |
Aspirator Bottle |
10 L | |
White | |
Closure, Handles | |
Narrow |