missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASPG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564429
This item is not returnable.
View return policy
Description
Immunogen sequence: VLVPGTGLAA ILRTLPMFHD EEHARARGLS EDTLVLPPAS RNQRILYTVL ECQPLFDSSD MTIAEWVCLA QTIKRHYEQY HGFVVIHGTD Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
Exhibits lysophospholipase, transacylase, PAF acetylhydrolase and asparaginase activities.
Specifications
| ASPG | |
| Polyclonal | |
| Unconjugated | |
| ASPG | |
| 1-alkyl-2-acetylglycerophosphocholine esterase; 60 kDa lysophospholipase; 60-kDa-lysophospholipase; asparaginase; asparaginase homolog; ASPG; C14orf76; GPA/WT; L-asparaginase; L-asparagine amidohydrolase; LysoLP; Lysophospholipase-transacylase; PAF acetylhydrolase; Platelet-activating factor acetylhydrolase; wild-type L-asparaginase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 374569 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86U10 | |
| ASPG | |
| Recombinant protein corresponding to Human ASPG. Recombinant protein control fragment (Product #RP-103960). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction