Learn More
Gilson™ ASPEC™ Alumina-N SPE Cartridge, Bed Wt. 1 g
Alumina cartridges contain a high-quality, polar sorbent, used the same way as silica, but with better pH stability (6 to 7.5), neutral character provides good retention of aromatic compounds, aliphatic amines and compounds with electronegative functions
Brand:  Gilson™ 54350778
Product Code. 16686455
  • £282.00 / Pack of 50
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Specifications
Specifications
Alumina-N
1 g
Alumina-N
6 mL
SPE
ASPEC™
SPE cartridge
Yes