missing translation for 'onlineSavingsMsg'
Learn More

Aspartyl-alanyl-diketopiperazine, MedChemExpress

Product Code. 16426807
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16426807

Brand: Medchem Express HY107091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Aspartyl-alanyl-diketopiperazine (DA-DKP) is an immunomodulatory molecule generated by cleavage and cyclization from the N-terminus of human albumin and can modulate the inflammatory immune response through a molecular pathway implicated in T- lymphocyte anergy.

Aspartyl-alanyl-diketopiperazine (DA-DKP) is an immunomodulatory molecule generated by cleavage and cyclization from the N-terminus of human albumin and can modulate the inflammatory immune response through a molecular pathway implicated in T- lymphocyte anergy.

Specifications

Chemical Name or Material Aspartyl-alanyl-diketopiperazine
CAS 110954-19-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H10N2O4
Quantity 5 mg
Synonym DA-DKP
Solubility Information DMSO : 125 mg/mL (671.43 mM; Need ultrasonic)
SMILES O=C(O)C[C@@H](C(N[C@H]1C)=O)NC1=O
Molecular Weight (g/mol) 186.17
Formula Weight 186.17
Percent Purity 99.44%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.