missing translation for 'onlineSavingsMsg'
Learn More

Aspartyl-alanyl-diketopiperazine, MedChemExpress

Product Code. 16494098
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494098 10 mg 10mg
16470457 50 mg 50mg
16426807 5 mg 5mg
16415076 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494098 Supplier Medchem Express Supplier No. HY107091
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aspartyl-alanyl-diketopiperazine (DA-DKP) is an immunomodulatory molecule generated by cleavage and cyclization from the N-terminus of human albumin and can modulate the inflammatory immune response through a molecular pathway implicated in T- lymphocyte anergy.

Aspartyl-alanyl-diketopiperazine (DA-DKP) is an immunomodulatory molecule generated by cleavage and cyclization from the N-terminus of human albumin and can modulate the inflammatory immune response through a molecular pathway implicated in T- lymphocyte anergy.

Specifications

Chemical Name or Material Aspartyl-alanyl-diketopiperazine
CAS 110954-19-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H10N2O4
Quantity 10 mg
Synonym DA-DKP
Solubility Information DMSO : 125 mg/mL (671.43 mM; Need ultrasonic)
SMILES O=C(O)C[C@@H](C(N[C@H]1C)=O)NC1=O
Molecular Weight (g/mol) 186.17
Formula Weight 186.17
Percent Purity 99.44%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.