missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aspartoacylase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529180
This item is not returnable.
View return policy
Description
Recommended positive controls: mouse brain. Predicted reactivity: Mouse (86%), Rat (86%), Pig (92%), Rhesus Monkey (96%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ASPA is an enzyme that catalyzes the conversion of N-acetyl_L-aspartic acid (NAA) to aspartate and acetate. NAA is abundant in the brain where hydrolysis by aspartoacylase is thought to help maintain white matter. This protein is an NAA scavenger in other tissues.
Specifications
| Aspartoacylase | |
| Polyclonal | |
| Unconjugated | |
| ASPA | |
| ACY2; ACY-2; aminoacylase 2; aminoacylase-2; ASP; ASPA; Aspartoacylase; aspartoacylase (aminoacylase) 2; nur7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11484, 443 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P45381, Q8R3P0 | |
| ASPA | |
| Full length human Aspartoacylase Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction