missing translation for 'onlineSavingsMsg'
Learn More

(-)-Aspartic acid, MedChemExpress

Product Code. 16430518
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16430518 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16430518 Supplier Medchem Express Supplier No. HY42068
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (-)-Aspartic acid is an endogenous NMDA receptor agonist.

(-)-Aspartic acid is an endogenous NMDA receptor agonist.

Specifications

Chemical Name or Material (-)-Aspartic acid
CAS 1783-96-6
Color White
Purity Grade Notes Research
Molecular Formula C4H7NO4
Synonym (R)-Aspartic acid D-(-)-Aspartic acid
Solubility Information H2O : 7.69 mg/mL (57.78 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (7.51 mM; Need ultrasonic)
SMILES N[C@@H](C(O)=O)CC(O)=O
Molecular Weight (g/mol) 133.1
Quantity 5 g
Formula Weight 133.1
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.