missing translation for 'onlineSavingsMsg'
Learn More

Aspartic acid calcium, MedChemExpress

Product Code. 16402549
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402549 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402549 Supplier Medchem Express Supplier No. HYN0666B
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Aspartic acid calcium (Calcium L-aspartate) is a chelate where calcium is attached to an amino acid naming L-Aspartic acid. L-Aspartic acid is an amino acid and serves as a building block for proteins in the body.

Aspartic acid calcium (Calcium L-aspartate) is a chelate where calcium is attached to an amino acid naming L-Aspartic acid. L-Aspartic acid is an amino acid and serves as a building block for proteins in the body.

Specifications

Chemical Name or Material Aspartic acid calcium
CAS 10389-09-0
Color White
Purity Grade Notes Research
Molecular Formula C4H5CaNO4
Synonym Calcium L-aspartate
Solubility Information H2O : ≥ 100 mg/mL (584.25 mM)
SMILES N[C@@H](CC([O-])=O)C([O-])=O.[Ca+2]
Molecular Weight (g/mol) 171.16
Quantity 1 g
Formula Weight 171.16
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.