missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aspartic Acid (Aspartic Acid), LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Aspartic Acid (Aspartic Acid)
Specifications
Specifications
| Chemical Name or Material | L-Aspartic acid |
| CAS | 56-84-8 |
| Analyte or Component Names | L-Aspartic acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H7 N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Aspartic Acid, L-Aspartic acid, Aspartic acid, L-, (+)-Aspartic acid, (S)-2-Aminobutanedioic acid, (S)-Aminobutanedioic acid, (S)-Aspartic acid, Asparagic acid, Asparaginic acid, Aspartic acid, Butanedioic acid, amino-, (S)-, H-Asp-OH, L-(+)-Aspartic acid, L-Aminosuccinic acid, L-Asparagic acid, L-Asparaginic acid, NSC 3973, NSC 79553, Alanine Impurity A, Asparagine Monohydrate Impurity A, Asparagine Impurity A |
| InChI Formula | InChI=1 S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1 |
| SMILES | N[C@@H](CC(=O)O)C(=O)O |
| IUPAC Name | (2 S)-2-aminobutanedioic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction